InLab® ORP Electrodes
Precision ORP probes to monitor oxidation-reduction reactions
  • Cmbination ORP electrodes
The ARGENTHAL™ reference system is silver free—the junction won't become blocked by sulfides or proteins.
144,000.00 - ₹222,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Electrode Type
Availability Pricing
Mettler Toledo Redox ORP Electrode; S7 3590230 Mfr # 51343200
COMPARE
Platinum
₹144,000.00INR / Each
Mettler Toledo Redox-L ORP Electrode; S7 3590231 Mfr # 51343202
COMPARE
Platinum
₹156,000.00INR / Each
Mettler Toledo Redox Flow ORP Electrode; S7 3590232 Mfr # 51343201
COMPARE
Platinum
₹222,000.00INR / Each
Mettler Toledo Redox Micro ORP Electrode; S7 3590233 Mfr # 51343203
COMPARE
Platinum
₹163,000.00INR / Each
Mettler Toledo Redox Ag ORP Electrode; S7 3590235 Mfr # 51343205
COMPARE
Silver
₹156,000.00INR / Each