Oakton Buffer Solution, pH 4.01; 1000 mL

Representative image only.
Oakton Item # 0594222
Calibrate pH instruments to ensure accuracy
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 4.01 traceable to NIST standard reference materials

PRODUCT OPTIONS

₹2,800.00INR / Each
Choose the buffer value and volume that fits your needs
Oakton® pH Buffer Solutions
2,000.00 - ₹31,900.00INR / Case of 4

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value4.01
  • Volume (mL)1000
  • ColorRed

Key Features

  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 4.01 traceable to NIST standard reference materials

More About this Item

pH 4 is a common low-range pH meter calibration point. All Oakton pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.

Available as Reference Materials (RM) and Certified Reference Material (CRM) grade. pH 4 buffer solution is colored red for visual reference. It is sold in individual 1000-mL (1-L) bottles. If unopened, shelf life of pH 4 is 2 years; once opened, use within 90 days.

Store buffers away from direct sunlight.

Store buffers away from direct sunlight.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat