Oakton Buffer Solution, pH 4.01; 4 x 1 gal. Bottles/Cs

Representative image only.
Oakton Item # 0594225
Calibrate pH instruments to ensure accuracy
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 4.01

PRODUCT OPTIONS

₹22,000.00INR / Case of 4
Choose the buffer value and volume that fits your needs
Oakton® pH Buffer Solutions
2,000.00 - ₹31,900.00INR / Case of 4

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value4.01
  • Volume (mL)4000
  • ColorRed

Key Features

  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 4.01

More About this Item

Standardized against NIST-traceable pH references for meters with 0.01 resolution. Sold as a case of four jugs; 1-gallon bottles are also available individually.

Store buffers away from direct sunlight.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat