Oakton Buffer Solution, pH 9.18; 5 x 60 mL Bottles/Pk

Representative image only.
Oakton Item # 0065312
Calibrate pH instruments to ensure accuracy
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST buffer value of pH 9.18
  • 1 year warranty

PRODUCT OPTIONS

₹21,300.00INR / Pkg of 5
Buffer Solutions…
Oakton® pH Buffer Solutions
2,000.00 - ₹31,900.00INR / Case of 4

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value9.18
  • Volume (mL)60
  • ColorClear
  • DescriptionBuffer Solution, pH 9.18; 5 x 60 mL Bottles/Pk
  • Warranty1 year
1 Year Warranty

Key Features

  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST buffer value of pH 9.18
  • 1 year warranty

More About this Item

Convenient 60-mL bottles come in a pack of five—ideal for field use. Grab one when you're on the go! Standardized against NIST-traceable pH references for meters with 0.01 resolution.

Store buffers away from direct sunlight.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat