Oakton Ion 6+ Handheld Meter with ATC Probe

Representative image only.
Oakton Item # 3561380
Direct readout of ion concentration at an economical price
  • Compact and easy-to-use with preprogrammed ion calibration points
  • mV mode allows you to check electrode calibration and performance
  • +/- 1999 mV range is compatible with virtually all ISE electrodes
  • 3 year warranty

PRODUCT OPTIONS

₹107,000.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Electrodes Today!

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01 or 0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionDIN, NIST, and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution0.1 or 1 mV
  • mV Accuracy±1mV + 1 digit
  • Min ISE0.01 to 1999 ppm
  • ISE Resolution0.01 ppm
  • ISE Accuracy±0.5% of reading
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Temperature compensationAutomatic
  • Battery4 AAA (included)
  • Battery life (hr)700
  • Width (cm)8.5725
  • Height (cm)4.28625
  • Width (in)3 3/8
  • Height (in)1 11/16
  • DescriptionHandheld Meter with ATC Probe
  • Warranty3 year
3 Year WarrantyConformite EuropeenneISO9001:2000 Certified Supplier

Includes

4 AAA batteries.

Key Features

  • Compact and easy-to-use with preprogrammed ion calibration points
  • mV mode allows you to check electrode calibration and performance
  • +/- 1999 mV range is compatible with virtually all ISE electrodes
  • 3 year warranty

More About this Item

These rugged, economical meters are very easy to use. Features include all push-button calibration, hold function, auto-off, and splash-resistant keypad. Read concentration in ppm and mV as well as pH and temperature in °C. The included protective rubber boot also acts as a meter stand for benchtop use. Order ISE electrodes separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat