Oakton® Ion 6+ Meters
Direct readout of ion concentration at an economical price
  • Compact and easy-to-use with preprogrammed ion calibration points
  • mV mode allows you to check electrode calibration and performance
  • +/- 1999 mV range is compatible with virtually all ISE electrodes
These rugged, economical meters are very easy to use. Features include all push-button calibration, hold function, auto-off, and splash-resistant keypad. Read concentration in ppm and mV as well as pH and temperature in °C. The included protective rubber boot also acts as a meter stand for benchtop use. Order ISE electrodes separately.
107,000.00 - ₹158,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Min ISE Description
Availability Pricing
Oakton Ion 6+ Handheld Meter with ATC Probe 3561380
₹107,000.00INR / Each
COMPARE
0.01 to 1999 ppm Handheld Meter with ATC Probe
Loading
₹107,000.00INR / Each
Oakton Ion 6+ Handheld Meter Kit w/ Case, Solutions, and pH/ATC Probe 3561382
₹135,000.00INR / Each
COMPARE
0.01 to 1999 ppm Handheld Meter Kit w/ Case, Solutions, and pH/ATC Probe
Loading
₹135,000.00INR / Each
Oakton Ion 6+ Handheld Meter Kit with NIST-Traceable Calibration 3561383
₹158,000.00INR / Each
COMPARE
0.01 to 1999 ppm Handheld Meter Kit with NIST-traceable Calibration
Loading
₹158,000.00INR / Each