Oakton pH 6+ Handheld Meter with pH Probe

Representative image only.
Oakton Item # 3561322
High +/-0.01 pH accuracy at an economical price
  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH or mV and temperature with the press of a button
  • 3 year warranty

PRODUCT OPTIONS

₹71,200.00INR / Each

₹25,500.00 - ₹34,500.00INR / Each

Time to recalibrate? Click on the calibration service to view details, then order.
Include Buffers Today!
Oakton® pH Buffer Solution Calibration Packs
5,100.00 - ₹51,000.00INR / Pkg of 3

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)0
  • Max pH (pH)14
  • pH Resolution0.01 or 0.1
  • pH Accuracy±0.01 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 5 points
  • Min mV (mV)-1999
  • Max mV (mV)1999
  • mV Resolution1 mV
  • mV Accuracy±0.2 mV or 0.05%, whichever is greater
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Temperature compensationAutomatic
  • Battery4 AAA (included)
  • Battery life (hr)700
  • Width (cm)8.5725
  • Height (cm)4.28625
  • Width (in)3 3/8
  • Height (in)1 11/16
  • Warranty3 year
3 Year WarrantyConformite Europeenne

Includes

35613-05 ATC probe, 59001-65 pH probe, protective rubber boot, and 4 AAA batteries.

Key Features

  • Convenient, compact design
  • Up to five-point push-button calibration with autobuffer recognition
  • Displays pH or mV and temperature with the press of a button
  • 3 year warranty

More About this Item

These meters are perfect for fast, accurate results. Automatic temperature compensation (ATC)ensures accuracy from 0 to 100°C. Meters include a protective rubber boot that also acts as a meter stand.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat