Oakton Traceable® pH CRM Buffer, Blue, pH 10; 500 mL

Representative image only.
Oakton Item # 0065110
pH buffers tested and certified for laboratory compliance
  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • NIST/USA buffer value of pH 10.001 traceable to NIST standard reference materials

PRODUCT OPTIONS

₹9,300.00INR / Each
Buffer Solutions…
Oakton® Traceable® CRM pH Buffer Solutions
3,600.00 - ₹28,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value10.001
  • Volume (mL)500
  • ColorBlue
NIST Traceable Calibration

Key Features

  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • NIST/USA buffer value of pH 10.001 traceable to NIST standard reference materials

More About this Item

Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with A2LA ISO/IEC 17025:2005, ISO Guide 34, ISO 31, and ISO 35, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express Oakton monitors the solution over 1 to 2 years and informs users of any changes.

pH 10 is a common high-range pH meter calibration point. All Oakton pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations. Available as Reference Materials (RM) and Certified Reference Material (CRM) grade. pH 10 buffer solution is available colored blue for visual reference or clear. It is sold in individual 500-mL bottles. If unopened, shelf life of pH 10 is 2 years; once opened, use within 30 days. Store buffers away from direct sunlight.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat