Oakton® Traceable® CRM pH Buffer Solutions

pH buffers tested and certified for laboratory compliance
  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with ISO/IEC 17025:2017, ISO 17034:2016, ISO Guide 31, and ISO 33405, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express Oakton monitors the solution over 1 to 2 years and informs users of any changes.
3,600.00 - ₹28,000.00INR / Each

19 variations of this product are available.

See All Product Options Below
19 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Value Color Volume (mL)
Availability Pricing
COMPARE
4.005 Clear 500
₹6,600.00INR / Each
COMPARE
7 Clear 500
₹6,500.00INR / Each
COMPARE
10.001 Clear 500
₹6,600.00INR / Each
COMPARE
4.005 Red 500
₹6,100.00INR / Each
COMPARE
7 Yellow 500
₹9,300.00INR / Each
COMPARE
10.001 Blue 500
₹9,300.00INR / Each
COMPARE
4.005 Clear 1000
₹6,600.00INR / Each
COMPARE
7 Clear 1000
₹6,600.00INR / Each
COMPARE
10.001 Clear 1000
₹6,600.00INR / Each
COMPARE
4.005 Red 1000
₹14,800.00INR / Each
COMPARE
7 Yellow 1000
₹14,800.00INR / Each
COMPARE
10.005 Blue 1000
₹14,800.00INR / Each
COMPARE
4.005 Clear 4000
₹13,100.00INR / Each
COMPARE
7 Clear 4000
₹13,100.00INR / Each
COMPARE
10.005 Clear 4000
₹13,400.00INR / Each
COMPARE
4.005 Red 4000
₹28,000.00INR / Each
COMPARE
7 Yellow 4000
₹28,000.00INR / Each
COMPARE
10.001 Blue 4000
₹28,000.00INR / Each
COMPARE
7.416 Clear 500
₹3,600.00INR / Each
Showing 1- 19 of 19

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat