Oakton Traceable® pH Standard Buffer with Calibration, Red, pH 4; 1000 mL

Representative image only.
Oakton Item # 0065136
pH buffers tested and certified for laboratory compliance
  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • 1000 mL (32 fl.oz.) capacity
  • Calibrate at pH 4
  • Easily identify red color-coded formula

PRODUCT OPTIONS

₹14,800.00INR / Each
Buffer Solutions…
Oakton® Traceable® CRM pH Buffer Solutions
3,600.00 - ₹28,000.00INR / Each

SPECS & DESCRIPTION

Specifications & Description

  • Buffer Value4.005
  • Volume (mL)1000
  • ColorRed
NIST Traceable Calibration

Key Features

  • Certified Reference Material for compliance-controlled laboratories and facilities
  • Available in multiple volumes to reduce waste and support various levels of lab throughput
  • Color-coded and/or clearly labeled for easy identification in busy labs
  • Supplied with calibration report from an ISO 17034 and ISO 17025 accredited laboratory
  • 1000 mL (32 fl.oz.) capacity
  • Calibrate at pH 4
  • Easily identify red color-coded formula

More About this Item

Certified Reference Materials (CRM) meet federal, state, and local requirements, providing standardized reference calibration for pH meters. Produced in compliance with A2LA ISO/IEC 17025:2005, ISO Guide 34, ISO 31, and ISO 35, CRMs ensure top-tier accuracy, documentation, and standards. They undergo extensive stability, transportation, and repeatability testing, with uncertainty (U) assigned from these evaluations. Calibration reports are issued by an ISO 17034 and ISO 17025 accredited lab. Environmental Express Oakton monitors the solution over 1 to 2 years and informs users of any changes.

Certificate of Analysis

Enter lot number
or click to view a Sample C of A

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat