Poly-Net 0400-YELLOW Heavy-gauge HDPE protective netting, 1-1/4" to 1-5/8" dia; 82 ft/pk

Representative image only.
Poly-Net Mfr # 0400-YELLOW – Item # 0940550
Minimize impact on glassware
  • Simply place over glassware to protect from bumps or flying debris
  • Cut to fit your needs
  • Constructed of HDPE
  • Yellow heavy-gauge protective netting

PRODUCT OPTIONS

₹32,500.00INR / Roll of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialHDPE; Fits 1-1/4 to 1-5/8" diameter
  • Thickness (in)0.11
  • Thickness (mm)2.8
  • Max Temperature (° F)175
  • Max Temperature (° C)80
  • Length (ft)82
  • Length (m)24.9936
  • ColorYellow

Key Features

  • Simply place over glassware to protect from bumps or flying debris
  • Cut to fit your needs
  • Constructed of HDPE
  • Yellow heavy-gauge protective netting

More About this Item

The heavy-gauge netting is more rigid for extra protection of your glassware. Different colored mesh options make for quick and easy visual indication.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat