Poly-Net™ Heavy-gauge Protective Netting

Minimize impact on glassware
  • Simply place over glassware to protect from bumps or flying debris
  • Cut to fit your needs
  • Constructed of HDPE
The heavy-gauge netting is more rigid for extra protection of your glassware. Different colored mesh options make for quick and easy visual indication.
32,500.00 - ₹45,500.00INR / Roll of 1

2 variations of this product are available.

See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Material
Availability Pricing
Poly-Net 0400-YELLOW Heavy-gauge HDPE protective netting, 1-1/4" to 1-5/8" dia; 82 ft/pk 0940550 Mfr # 0400-YELLOW
COMPARE
82 HDPE; Fits 1-1/4 to 1-5/8" diameter
₹32,500.00INR / Roll of 1
Poly-Net 0413-GREEN Heavy-gauge HDPE protective netting, 2-1/2" to 3" dia; 82 ft/pk 0940555 Mfr # 0413-GREEN
COMPARE
25 HDPE; Fits 2-1/2 to 3" diameter
₹45,500.00INR / Roll of 1
Showing 1- 2 of 2

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat