Sartorius Arium Mini Plus Water Purification System

Representative image only.
Sartorius Mfr # H2O-MA-T-US – Item # 9929320
Eliminate tank cleaning with unique closed bag system
  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Multicolor touch display with clear icons simplifies operation
  • Compact design with only a 11" (28 cm) wide footprint
  • 2 year warranty

PRODUCT OPTIONS

Call For Price

SPECS & DESCRIPTION

Specifications & Description

  • ASTM Water GradeType I & Type III
  • Max Flow Rate (L/min)1
  • Inlet Port0.25" OD tubing
  • With UV LampNo
  • With UltrafilterYes
  • SourceDirect conection to portable water supply
  • Type I Water (Resistivity)18.2 MΩ-cm
  • Feed Water Requirements (TOC)<2000 ppb
  • Type I Water (Conductivity)0.055 µS/cm
  • Type I Water (Bacteria)<1 CFU/1,000 mL
  • Feed Water Requirements (pressure)0.5 to 6 bar
  • Feed Water Requirements (Temperature)2 to 30°C
  • Max Inlet Water Temperature (° C)30
  • Min Inlet Water Temperature (° C)2
  • Max Inlet Pressure (PSI)6
  • Min Inlet Pressure (bar)0.5
  • Power (VAC)100 to 240
  • Height (in)20.06
  • Depth (in)20.89
  • Width (in)11.02
  • Height (cm)50.94
  • Depth (cm)53.07
  • Width (cm)28
  • DescriptionWater Purification System
  • Warranty2 year
Two year warranty

Key Features

  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Multicolor touch display with clear icons simplifies operation
  • Compact design with only a 11" (28 cm) wide footprint
  • 2 year warranty

More About this Item

The compact ultrapure water system features a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals. System features include a favorites function to dispense recurring volumes, integrated pump for increasing water pressure, and pretreatment cartridge for effective purification.

Two models are available. The arium mini version is manually fed with pretreated water, while the arium mini plus version has direct connection to the portable water supply. Both are available with UV lamp (185/254 nm) to make sure the water is virtually free of organic components.

Consumables are needed to operate the unit and are sold separately. Please purchase a starter set along with your water system.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat