Sartorius Arium® Mini and Mini Plus Water Systems
Eliminate tank cleaning with unique closed bag system
  • Ideal for small labs requiring <10 L/day of ultrapure water
  • Multicolor touch display with clear icons simplifies operation
  • Compact design with only a 11" (28 cm) wide footprint
The compact ultrapure water system features a closed bag tank system which protects pure water from secondary contamination and ensures consistent long-term high water quality. The 5-L bag can be quickly and easily replaced every six months, reducing maintenance time and eliminating the use of hazardous cleaning chemicals. System features include a favorites function to dispense recurring volumes, integrated pump for increasing water pressure, and pretreatment cartridge for effective purification.
Two models are available. The arium mini version is manually fed with pretreated water, while the arium mini plus version has direct connection to the portable water supply. Both are available with UV lamp (185/254 nm) to make sure the water is virtually free of organic components.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ASTM Water Grade Source With UV Lamp
Availability Pricing
Sartorius Arium Mini Water Purification System 9929310 Mfr # H2O-MM-T-US
COMPARE
Type I Manual feeed with pre-treated water No
Call For Price
Sartorius Arium Mini Water Purification System with UV lamp 9929312 Mfr # H2O-MM-UV-T-US
COMPARE
Type I Manual feeed with pre-treated water Yes
Call For Price
Sartorius Arium Mini Plus Water Purification System 9929320 Mfr # H2O-MA-T-US
COMPARE
Type I & Type III Direct conection to portable water supply No
Call For Price
Sartorius Arium Mini Plus Water Purification System with UV lamp 9929322 Mfr # H2O-MA-UV-T-US
COMPARE
Type I & Type III Direct conection to portable water supply Yes