Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 45mm Flat Width, 28.6mm dia; 100' L

Representative image only.
Spectra Por Mfr # 132706 – Item # 0290044
Get gentle separation and purification of solutes
  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000 daltons
  • 6 month warranty

PRODUCT OPTIONS

₹75,200.00INR / Box of 1

SPECS & DESCRIPTION

Specifications & Description

  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Flat Width (mm)45
  • Diameter (mm)28.6
  • Length Per Pack (ft)100
  • Diameter (in)1.126
  • Description4 Membranes; flat width, 45mm; diameter, 28.6mm; length, 100'
  • Warranty6 month
6 Month Warranty

Key Features

  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000 daltons
  • 6 month warranty

More About this Item

The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 and 30 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
Similar to SPECTRA/POR 2 tubing, except thicker. Dialysis time is slower, but at much less cost.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat