Spectra/Por 4 Regenerated Cellulose (RC) Dialysis Tubing
Get gentle separation and purification of solutes
  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000 daltons
The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 and 30 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
Similar to SPECTRA/POR 2 tubing, except thicker. Dialysis time is slower, but at much less cost.
60,600.00 - ₹104,000.00INR / Box of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Flat Width (mm) Max Molecular Weight Cut Off (Mwco) (Dalton)
Availability Pricing
Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 10mm Flat Width, 6.4mm dia; 100' L 0290041 Mfr # 132697
COMPARE
6.4 10 14000
₹104,000.00INR / Box of 1
Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 25mm Flat Width, 15.9mm dia; 100' L 0290042 Mfr # 132700
COMPARE
15.9 25 14000
₹60,600.00INR / Box of 1
Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 32mm Flat Width, 20.4mm dia; 100' L 0290043 Mfr # 132703
COMPARE
20.4 32 14000
₹64,900.00INR / Box of 1
Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 45mm Flat Width, 28.6mm dia; 100' L 0290044 Mfr # 132706
COMPARE
28.6 45 14000
₹75,200.00INR / Box of 1
Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 75mm Flat Width, 47.7mm; dia; 50' L 0290045 Mfr # 132709
COMPARE
47.7 75 14000
₹78,700.00INR / Box of 1