Thermo Scientific Nunc 534592 Miniboxes For 1.0 - 1.8 mL Cryotubes, 200/pk

Representative image only.
Thermo Scientific Nunc Mfr # 534592 – Item # 0375509
Secure storage and shipping of valuable cryogenic samples
  • Friction-fit lids keep vials securely in place
  • Durable high-impact polystyrene construction
  • Available in two sizes to hold 1.0 and 2.0 mL vials

PRODUCT OPTIONS

₹272,000.00INR / Pkg of 200

SPECS & DESCRIPTION

Specifications & Description

  • Tubes Held10
  • Tube Size1.0 to 1.8 mL
  • ColorWhite
  • DescriptionMiniboxes For 1.0 - 1.8 mL Cryotubes, 200/pk

Key Features

  • Friction-fit lids keep vials securely in place
  • Durable high-impact polystyrene construction
  • Available in two sizes to hold 1.0 and 2.0 mL vials

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat