Thermo Scientific Nunc CryoTube Mini Boxes
Secure storage and shipping of valuable cryogenic samples
  • Friction-fit lids keep vials securely in place
  • Durable high-impact polystyrene construction
  • Available in two sizes to hold 1.0 and 2.0 mL vials
272,000.00 - ₹367,000.00INR / Pkg of 50
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Tube Size
Availability Pricing
Thermo Scientific Nunc 534479 Miniboxes For 1.0 - 1.8 mL Cryotubes, 350/pk 0375508 Mfr # 534479
COMPARE
5 1.0 to 1.8 mL
₹367,000.00INR / Pkg of 50
Thermo Scientific Nunc 534592 Miniboxes For 1.0 - 1.8 mL Cryotubes, 200/pk 0375509 Mfr # 534592
COMPARE
10 1.0 to 1.8 mL
₹272,000.00INR / Pkg of 200