Thermo Scientific Nunc Microplate Cap Mat, 96-Well, TPE, Sterile; 50/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 276000 – Item # 0192954
Secure samples within your 96-well microplates
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
  • 96-well caps for plates 01929-32 and -33
PRODUCT OPTIONS
₹120,000.00INR / Case of 50
Often Bought With…
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well ShapeRound
  • Surface TreatmentNone
  • SterileYes
  • Plate TypeCap Mat
  • Plate ColorNatural
  • Min Temperature (° C)-20
  • Max Temperature (° C)130
  • MaterialThermoplastic elastomers (TPE)
Key Features
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
  • 96-well caps for plates 01929-32 and -33
More About this Item
Choose from a range of options to prevent contamination and evaporation of well contents and safely store samples within your 96-well plates.
Lids have notices for quick orientation and have individual condensation rings for enhanced sample security.
Sealing tap is an adhesive seal that effectively seals all microplate formats. Pressure sensitive adhesive is not tacky to touch but forms a strong bond to the plate upon application of pressure.
Cap Mats create a tight seal but are easy to peel off without aerosols—reducing the risk of well-to-well contamination. Caps are pierceable by hypodermic needles without compromising seal.
Caps create a tight seal, but are easy to peel off without aerosols—reducing the risk of
well-to-well contamination. Caps are pierceable by hypodermic needles without
compromising seal.