Thermo Scientific Nunc MicroWell™ Microplates, 96-Well, Polypropylene, Non-Treated, Sterile, Round, 0.5 mL; 120/Cs
Representative image only.
Thermo Scientific Nunc Mfr # 267334 – Item # 0192933
Low binding polypropylene is ideal for homogeneous assays and storage
  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable
  • 1 year warranty
PRODUCT OPTIONS
₹172,000.00INR / Case of 120
Often Bought With…
Thermo Scientific™ Nunc™ Microplate Covers
61,200.00 - ₹120,000.00INR / Case of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.5
  • Max Working Well Volume (mL)0.45
  • Well ShapeRound
  • Surface TreatmentNon-Treated
  • Lid (Included)No
  • SterileYes
  • Plate ColorNatural
  • Min Temperature (° C)-80
  • Max Temperature (° C)121
  • MaterialPolypropylene
  • Width (mm)86
  • Length (mm)128
  • Warranty1 year
1 Year Warranty
Key Features
  • Shared wall design increases the well volume
  • Nontreated polypropylene plates are solvent resistant and autoclavable
  • 1 year warranty
More About this Item
Achieve complete sample recovery with the Thermo Scientific™ Nunc™ MicroWell™ 96-well microplates. Plates are ideal for storage as polypropylene has a lower binding capacity so proteins or DNA will not bind. Withstands temperatures from −80° to 121°C (−112° to 250°F).
Choose from conical bottom wells for optimal sample recovery or round bottom wells which are wider for improved mixing.