Thermo Scientific Nunc Microplate Lid, 96-Well, Polystyrene, Sterile; 50/Cs

Representative image only.
Thermo Scientific Nunc Mfr # 264122 – Item # 0192911
Secure samples within your 96-well microplates
  • Protect samples from contamination and evaporation during assay processing, incubation or storage
  • Lids for plates 01929-32, -33, -34, and -38

PRODUCT OPTIONS

₹61,200.00INR / Case of 50
Often Bought With…

SPECS & DESCRIPTION

Specifications & Description

  • Number of Wells96
  • Well ShapeAll Well Shapes
  • Surface TreatmentNone
  • SterileYes
  • Plate TypeLid
  • Plate ColorClear
  • Min Temperature (° C)-20
  • Max Temperature (° C)80
  • MaterialPolystyrene
  • Height (in)19/64
  • Width (in)3 11/32
  • Length (in)5
  • Height (mm)7.9
  • Width (mm)85
  • Length (mm)127
  • DescriptionMicroplate Lid, 96-Well, Polystyrene, Sterile; 50/Cs

Key Features

  • Protect samples from contamination and evaporation during assay processing, incubation or storage
  • Lids for plates 01929-32, -33, -34, and -38

More About this Item

Choose from a range of options to prevent contamination and evaporation of well contents and safely store samples within your 96-well plates.

Lids have notices for quick orientation and have individual condensation rings for enhanced sample security.

Sealing tap is an adhesive seal that effectively seals all microplate formats. Pressure sensitive adhesive is not tacky to touch but forms a strong bond to the plate upon application of pressure.

Cap Mats create a tight seal but are easy to peel off without aerosols—reducing the risk of well-to-well contamination. Caps are pierceable by hypodermic needles without compromising seal.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat