Torrey Pines EchoTherm HS30-600 Immersion Probe, 6" Stainless Steel, Platinum RTD, 3' Lead.
Representative image only.
Torrey Pines EchoTherm Mfr # HS30-600 – Item # 0464615
  • Compatible with EchoTherm digital and digital programmable hot plates and stirring hot plates
  • 1 year warranty
PRODUCT OPTIONS
₹32,800.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Warranty1 year
1 Year Warranty
Key Features
  • Compatible with EchoTherm digital and digital programmable hot plates and stirring hot plates
  • 1 year warranty
More About this Item
Use immersion probes to provide sample temperature feedback for more precise temperature control.