Immersion Probes for EchoTherm Stirring Hot Plates
  • Compatible with EchoTherm digital and digital programmable hot plates and stirring hot plates
Use immersion probes to provide sample temperature feedback for more precise temperature control.
18,600.00 - ₹38,500.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Torrey Pines EchoTherm HS30-600 Immersion Probe, 6" Stainless Steel, Platinum RTD, 3' Lead. 0464615 Mfr # HS30-600
COMPARE
Immersion Probe, 6" Stainless Steel, Platinum RTD, 3' Lead.
₹32,800.00INR / Each
Torrey Pines EchoTherm HS30-601 Immersion Probe, 10" Stainless Steel, Platinum RTD, 3' Lead. 0464620 Mfr # HS30-601
COMPARE
Immersion Probe, 10" Stainless Steel, Platinum RTD, 3' Lead.
₹38,500.00INR / Each
Torrey Pines EchoTherm HS30-602 Immersion Probe, 6" PTFE 0464625 Mfr # HS30-602
COMPARE
Stainless Steel Immersion Temperature Probe, 6" Length
₹36,200.00INR / Each
Torrey Pines EchoTherm HS30-700 Temperature Probe Calibration Kit 0464635 Mfr # HS30-700
COMPARE
Temperature Probe Calibration Kit
₹18,600.00INR / Each