Troemner WA600 Microfibre Cleaning Cloth
Representative image only.
Troemner Mfr # WA600 – Item # 0102814
  • Brushes and cloth are designed to remove visible particles from your weights.
  • Small 100 mm brushes are used for weights 1 mg through 500 g. Wide, medium, large and extra large brushes are designed for weights 1 kg through 5 kg. The brushes can be used on all weight styles, except Cast Iron.
  • The microfibre cloth is recommended for use on stainless steel weights only.
PRODUCT OPTIONS
₹3,400.00INR / Each
SPECS & DESCRIPTION
Specifications & Description
  • Mass CertificationNot applicable
  • DescriptionMicrofibre Cleaning Cloth
Key Features
  • Brushes and cloth are designed to remove visible particles from your weights.
  • Small 100 mm brushes are used for weights 1 mg through 500 g. Wide, medium, large and extra large brushes are designed for weights 1 kg through 5 kg. The brushes can be used on all weight styles, except Cast Iron.
  • The microfibre cloth is recommended for use on stainless steel weights only.