Troemner Mass Accessories - Brushes and Coth
  • Brushes and cloth are designed to remove visible particles from your weights.
  • Small 100 mm brushes are used for weights 1 mg through 500 g. Wide, medium, large and extra large brushes are designed for weights 1 kg through 5 kg. The brushes can be used on all weight styles, except Cast Iron.
  • The microfibre cloth is recommended for use on stainless steel weights only.
3,400.00 - ₹3,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Troemner WA195 100 mm Brush - 5 mm Diameter Brush Head 0102808 Mfr # WA195
COMPARE
100 mm Brush - 5 mm Diameter Brush Head
₹3,800.00INR / Each
Troemner WA600 Microfibre Cleaning Cloth 0102814 Mfr # WA600
COMPARE
Microfibre Cleaning Cloth
₹3,400.00INR / Each