Zefon Cassette, 37 mm, 2 Piece, 1.0 µm, PTFE with Polypropylene Support; 50/Bx

Representative image only.
Zefon Item # 6006102
Fully assembled and preloaded with PTFE membrane
  • Leak-free design—eliminates sample loss
  • Preloaded with 37-mm diameter PTFE membrane
  • Clear styrene housing provides excellent visibility of sample
  • Conform to most NIOSH standards
  • 1.0 µm PTFE membrane with PP support
  • Two-piece, clear styrene housing

PRODUCT OPTIONS

₹72,700.00INR / Box of 50

SPECS & DESCRIPTION

Specifications & Description

  • Filter Diameter (mm)37
  • Pore Size (µm)1
  • Body MaterialStyrene
  • SupportPolypropylene
  • DescriptionCassette, Preloaded, 37 mm, 2 Piece, Styrene, 1.0 µm, PTFE with PP Support; 50/BX

Key Features

  • Leak-free design—eliminates sample loss
  • Preloaded with 37-mm diameter PTFE membrane
  • Clear styrene housing provides excellent visibility of sample
  • Conform to most NIOSH standards
  • 1.0 µm PTFE membrane with PP support
  • Two-piece, clear styrene housing

More About this Item

Make air sampling fast and simple with preassembled, preloaded filter cassettes. Cassettes come preloaded with chemical-resistant PTFE membrane filter, making them ideal for sampling in harsh environments. Select membranes with polypropylene (PP) support for added durability; unsupported membrane for NIOSH-method 5013 testing. Cassettes are preloaded in an ISO 9001:2015-certified facility ensuring high quality control standards.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat