Zefon Preloaded Air Sampling Cassettes with PTFE Filter Membrane
Fully assembled and preloaded with PTFE membrane
  • Leak-free design—eliminates sample loss
  • Preloaded with 37-mm diameter PTFE membrane
  • Clear styrene housing provides excellent visibility of sample
  • Conform to most NIOSH standards
Make air sampling fast and simple with preassembled, preloaded filter cassettes. Cassettes come preloaded with chemical-resistant PTFE membrane filter, making them ideal for sampling in harsh environments. Select membranes with polypropylene (PP) support for added durability; unsupported membrane for NIOSH-method 5013 testing. Cassettes are preloaded in an ISO 9001:2015-certified facility ensuring high quality control standards.
40,100.00 - ₹85,200.00INR / Box of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Pore Size (µm) Support Description
Availability Pricing
COMPARE
1 Polypropylene
₹72,700.00INR / Box of 50
COMPARE
1 Polypropylene
₹83,100.00INR / Box of 50
COMPARE
0.45 Polypropylene
₹85,200.00INR / Box of 50
COMPARE
0.5 PTFE
₹40,100.00INR / Box of 50