Bausch & Lomb Measuring Scales
Improve inspection measurements
  • Glass measuring scales are compatible with measuring magnifiers
Use these measuring scales with the Hastings measuring magnifiers. Scales simply screw onto the bottom of each magnifier lens.
Measuring magnifiers 03882-10 and -50 are sold separately.
8,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Lens Magnification Description
Availability Pricing
Bausch & Lomb Measuring Scale, General Purpose Multi-Scale 0388211 Mfr # 81-34-36
COMPARE
Radii from 1/16" to 3/8", angles in single degrees to 90°, 0.005" and 0.1 mm scales, and line widths of 0.001", 0.002" and 0.003" None Measuring Scale, General Purpose Multi-Scale
₹8,700.00INR / Each
Bausch & Lomb Measuring Scale, Metric, 20 mm in 0.1 mm Increments 0388213 Mfr # 81-34-38
COMPARE
20 mm Scale with 0.1 mm increments None Measuring Scale, Metric, 20 mm in 0.1 mm Increments
₹8,700.00INR / Each