Binder Programmable Mechanical Convection Ovens
Microprocessor program controller allows easy programming of variable temperature and time functions
  • 5°°C above ambient to 300°C
  • Microprocessor controller with LED display and integrated 0 to 99-hr timer
  • APT.Line® preheating chamber ensures accurate, reproducible results
These programmable ovens offer electronically controlled APT.line® preheating chamber technology with forced air convection. Microprocessor with two programs of ten sections each, alternatively 1 program with 20 sections. The time interval of single program sections can be adjusted up to a maximum of 99 hours and 59 minutes, applicable to all program sections. Program controller functions: delayed ON, delayed OFF, temperature-dependent delayed OFF.
Additional oven features include adjustable ramp functions via program editor, adjustable fan speed (0 to 100%), elapsed time indicator, and adjustable safety device, Class 2 (DIN 12880), with visual alarm. Ventilation may be adjusted by means of a 50-mm diameter rear exhaust duct with ventilation flap and front side ventilation. RS-422 interface and APT-COM® communications software allow output to computer or printer. Ovens are stackable to conserve valuable lab space.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC)
Availability Pricing
Binder FP 53-UL FP Programmable Mechanical Convection Oven; 1.9 cu ft, RS-422, 115V 0501229 Mfr # 9010-0175
COMPARE
1.9 115
Call For Price
Binder 9010-0153 FP Programmable Mechanical Convection Oven; 1.9 cu ft, RS-422, 230V 0501231 Mfr # 9010-0153
COMPARE
2 230
Call For Price
Binder 9010-0262 FP Programmable Mechanical Convection Oven; 4.1 cu ft, RS-422, 115V 0501232 Mfr # 9010-0262
COMPARE
4 115
Call For Price
Binder 9010-0255 FP Programmable Mechanical Convection Oven; 4.1 cu ft, RS-422, 230V 0501233 Mfr # 9010-0255
COMPARE
4 230