Caframo High-Torque Digital Brushless Mixers
Chuck guard encloses spinning chuck for added safety
  • Set speed maintained as fluid viscosity changes
  • Includes tare function to set torque reading at zero
The unique, two-speed transmission delivers constant performance under changing condistions. Mixer features a sparkless, brushless DC motor that provides a long, maintenance-free service life. The totally enclosed non-ventilated motor is rated for continuous duty. Overload protection automatically shuts off the mixer if maximum torque is exceeded — an LED light signals overload conditions. The through-shaft design lets you easily alter shaft length and propeller depth. Mixers have an aluminum alloy housing.
458,000.00 - ₹479,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Speed (rpm) Max Torque (in-oz) Power (VAC)
Availability Pricing
Caframo High-Torque Brushless Mixer; 12 to 1800 rpm, 115 VAC 5080000 Mfr # BDC1850
COMPARE
1800 821 115
₹458,000.00INR / Each
Caframo High-Torque Brushless Mixer; 12 to 1800 rpm, 230 VAC 5080005 Mfr # BDC1850220U1
COMPARE
1800 821 230
₹479,000.00INR / Each
Caframo High-Torque Brushless Mixer; 20 to 3000 rpm, 115 VAC 5080010 Mfr # BDC3030
COMPARE
3000 496 115
₹458,000.00INR / Each
Caframo High-Torque Brushless Mixer; 20 to 3000 rpm, 220-240 VAC 5080015 Mfr # BDC3030220U1
COMPARE
3000 496 220-240
₹479,000.00INR / Each
Caframo High-Torque Brushless Mixer; 40 to 6000 rpm, 115 VAC 5080020 Mfr # BDC6015
COMPARE
6000 247 115
₹458,000.00INR / Each
Caframo High-Torque Brushless Mixer; 40 to 6000 rpm, 230 VAC 5080025 Mfr # BDC6015220U1
COMPARE
6000 247 230
₹479,000.00INR / Each