Cole-Parmer® Autoclavable Media Bottles, Square Plastic, PPCO
Autoclavable plastic media storage bottles for safe storage of your chemicals and media formulations
  • Resistant to acids, alcohols, and bases
  • Drip proof spout for easy pouring
  • Space-saving square design
  • Autoclavable
These translucent plastic media storage bottles feature molded-in mL graduations and a drip proof spout for easy pouring. The space saving, square shape helps you maximize shelf space. Made of polypropylene copolymer with a polypropylene cap, they are resistant to acids, alcohols, and bases, and can withstand temperatures from 0 to 135 °C (32 to 275 °F). These reusable media bottles are autoclavable for sterilization.
4,100.00 - ₹11,900.00INR / Pkg of 12
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Overall Height (cm)
Availability Pricing
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 30 mL (1 oz); 12/Pk 0625100
₹4,100.00INR / Pkg of 12
COMPARE
30 10 6.6
Loading
₹4,100.00INR / Pkg of 12
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 60 mL (2 oz); 12/Pk 0625110
₹4,200.00INR / Pkg of 12
COMPARE
60 10 8.2
Loading
₹4,200.00INR / Pkg of 12
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 125 mL (4.2 oz); 12/Pk 0625120
₹5,500.00INR / Pkg of 12
COMPARE
125 25 10.6
Loading
₹5,500.00INR / Pkg of 12
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 250 mL (8.5 oz); 12/Pk 0625130
₹8,400.00INR / Pkg of 12
COMPARE
250 25 14.1
Loading
₹8,400.00INR / Pkg of 12
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 500 mL (16.9 oz); 12/Pk 0625140
₹11,900.00INR / Pkg of 12
COMPARE
500 50 17.3
Loading
₹11,900.00INR / Pkg of 12
Cole-Parmer Autoclavable Media Bottles, Square Plastic, PPCO, 1000 mL (33.8 oz); 6/Pk 0625150
₹9,500.00INR / Pkg of 6
COMPARE
1000 50 21.6
Loading
₹9,500.00INR / Pkg of 6