Cole-Parmer® Ceramic Tweezers
Suitable for handling EOS/ESD sensitive components
  • Electrically insulated up to 932°F (500°C)
  • Highly corrosion resistant and nearly chemically inert
  • Lifetime Warranty
Full ceramic tweezers made from zirconia, are ideal for handling sensitive electronic components during thermal, chemical and soldering applications. The tweezers feature superior flexural strength, high temperature stability, and good abrasion and wear resistance.
56,400.00 - ₹57,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
ESD Safe Material Overall Length (in)
Availability Pricing
COMPARE
Yes Ceramic 4 1/2
₹57,300.00INR / Each
COMPARE
Yes Ceramic 4 59/64
₹57,300.00INR / Each
COMPARE
Yes Ceramic 4 59/64
₹56,400.00INR / Each
COMPARE
Yes Ceramic 4 59/64
₹56,400.00INR / Each