Cole-Parmer® Crucibles, PTFE Flouropolymer
Chemically inert PTFE crucibles have machine-finished surfaces that allow easy cleaning and help eliminate contamination. Withstand temperatures to 536°F (280°C). Cover is not included.
4,200.00 - ₹11,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Overall Height (mm) Capacity (mL) Top Outside Diameter (mm)
Availability Pricing
COMPARE
31 5 16
₹4,200.00INR / Each
COMPARE
44 25 26
₹6,100.00INR / Each
COMPARE
56 75 41
₹10,000.00INR / Each
COMPARE
46 100 73
₹11,600.00INR / Each