Cole-Parmer® Economical In-Line pH Electrodes
Integral, one-piece electrode and housing for installation convenience
  • Choose from single-junction or double-junction designs
  • Options of BNC or stripped and tinned ends
In-line electrodes operate from 32 to 212°F (0 to 100°C) and have a chemical-resistant PVDF housing with a 1/2'' NPT(M) thread. Housing measures 4'' x 15/16'' diameter. Electrode extends 1'' beyond housing. Maximum pressure 100 psi. Measure pH from 0 to 13.
23,200.00 - ₹67,700.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Cole-Parmer In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Tinned/Tinned 0599338
₹33,800.00INR / Each
COMPARE
In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Tinned/Tinned
Loading
₹33,800.00INR / Each
Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned 0599348
₹31,900.00INR / Each
COMPARE
In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/Tinned
Loading
₹31,900.00INR / Each
Oakton by Cole-Parmer® In-Line pH Probe, SJ/PVDF/100Ohm RTD/10'; Plug/BNC 0599370
₹26,600.00INR / Each
COMPARE
In-Line pH Probe, SJ/PVDF/100Ohm RTD/10'; Plug/BNC
Loading
₹26,600.00INR / Each
Cole-Parmer In-Line pH Probe, SJ/PVDF/100Ohm RTD/10'; Tinned/BNC 0599379
₹26,600.00INR / Each
COMPARE
In-Line pH Probe, SJ/PVDF/100Ohm RTD/10'; Tinned/BNC
Loading
₹26,600.00INR / Each
Oakton by Cole-Parmer® pH electrode, in-line, single-junction, no ATC, 10-ft cable, BNC 0599390
₹23,200.00INR / Each
COMPARE
pH electrode, in-line, single-junction, no ATC, 10-ft cable, BNC
Loading
₹23,200.00INR / Each
Oakton by Cole-Parmer® In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Plug/BNC 2700170
₹32,200.00INR / Each
COMPARE
In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Plug/BNC
Loading
₹32,200.00INR / Each
Oakton by Cole-Parmer® In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Tinned/BNC 2700172
₹31,600.00INR / Each
COMPARE
In-Line pH Probe, DJ/PVDF/100Ohm RTD/10'; Tinned/BNC
Loading
₹31,600.00INR / Each
Oakton pH electrode, in-line, double-junction, no ATC 10-ft cable, BNC 2700190
₹25,500.00INR / Each
COMPARE
pH electrode, in-line, double-junction, no ATC 10-ft cable, BNC
Loading
₹25,500.00INR / Each
Cole-Parmer In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/BNC 2700191
₹67,700.00INR / Each
COMPARE
In-Line pH Probe, DJ/PVDF/1kOhm RTD/10'; Spade/BNC
Loading
₹67,700.00INR / Each
Showing 1- 9 of 9