Cole-Parmer® Microscopy Fiber Optic Illuminators
Designed specifically for your microscope needs
  • Low profile design fits into tight spaces
  • Complete 0 to 100% light intensity control
These high-performance illuminators deliver 400,000 footcandles of high-intensity cold light. Minimal light leakage makes these illuminators ideal for your photosensitive applications.
Audible noise and vibration is minimal, convenient side door makes changing lamps fast and easy, light pipes fit securely without need for adapters. Built-in handle stays out of the way.
183,000.00 - ₹282,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Power (VAC) Description
Availability Pricing
COMPARE
230 Microscopy Fiber Optic Illuminator, No Light Pipe; 230 VAC
₹183,000.00INR / Each
COMPARE
230 Microscopy Fiber Optic Illuminator w/Gooseneck; 230 VAC
₹246,000.00INR / Each
COMPARE
230 Microscopy Fiber Optic Illuminator w/Dual Gooseneck; 230 VAC
₹282,000.00INR / Each