Cole-Parmer® Narrow-Mouth Plastic Bottle, PFA
Provides safe storage of corrosive and hazardous chemicals
  • Withstands temperatures from -200℃ to 260℃ (-328° to 500°F)
  • Chemical and corrosion resistance
  • Smooth inner and outer surface for easy cleaning
  • Autoclavable
These perfluoroalkoxy (PFA) narrow-mouth offer excellent resistance to chemicals, high and low temperatures, and corrosion. Recommended for demanding fluid handling and storage applications. Safe for reagents which can’t be stored in metal containers.
The translucent bottles can also be used for solutions or chemicals commonly used for trace quantity analysis, ultra-trace quantity analysis, ICP-MS analysis, and isotopic analysis. They feature extremely low water-absorption, leakproof PFA caps, and non-stick surface for easy cleaning.
9,600.00 - ₹26,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Neck Inside Diameter (ID) (mm)
Availability Pricing
COMPARE
60 18 mm, OD 24 mm 14
₹9,600.00INR / Each
COMPARE
100 26.6 mm, OD 36 mm 22
₹11,800.00INR / Each
COMPARE
250 GL32 25
₹16,000.00INR / Each
COMPARE
500 GL32 25
₹26,000.00INR / Each