Cole-Parmer® Narrow-Mouth Plastic Bottles, FEP
Maintain sterility of samples
  • Withstands repeated sterilization
  • Use in a wide temperature range from –328 to 401°F (–200 to 205°C)
  • Autoclavable
These FEP bottles can withstand repeated sterilization at temperatures from –328 to 401°F (–200 to 205°C). Use them for cryogenic experiments, trace metal analysis, and organic solvents.
32,000.00 - ₹89,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap / Stopper Size Material
Availability Pricing
COMPARE
125 GL25 Fluorinated Ethylene Propylene (FEP)
₹32,000.00INR / Each
COMPARE
500 GL32 Fluorinated Ethylene Propylene (FEP)
₹63,000.00INR / Each
COMPARE
1000 GL32 Fluorinated Ethylene Propylene (FEP)
₹89,700.00INR / Each