Cole-Parmer® RTD Probe with Handle
Designed to give the same accuracy and benefits of a RTD Probe in a compact economical design
  • Time constant : 1-3 seconds or greater
  • Custom designs or changes to this product are available upon request
Maximum error for probe assembly is ±1.1°C + 0.12% of reading below 300°F, ±0.35% of reading above 300°F. Alpha coefficient is 0.003850 Ω/Ω/°C (DIN IEC 751). Resistance at ice point (0°C) is 100 Ω.
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
21,100.00 - ₹26,600.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Sheath Length (in)
Availability Pricing
COMPARE
Digi-Sense 3-pin 932 10
₹24,000.00INR / Each
COMPARE
Digi-Sense 3-pin 932 18
₹26,400.00INR / Each
COMPARE
Digi-Sense 3-pin 932 10
₹23,800.00INR / Each
COMPARE
Digi-Sense 3-pin 932 18
₹26,600.00INR / Each
COMPARE
Digi-Sense 3-pin 932 8.1
₹23,600.00INR / Each
COMPARE
Digi-Sense 3-pin 700 2
₹23,700.00INR / Each
COMPARE
Digi-Sense 3-pin 900 4
₹23,700.00INR / Each
COMPARE
Digi-Sense 3-pin 400 10
₹26,200.00INR / Each
COMPARE
Digi-Sense 3-pin 932 10
₹23,600.00INR / Each
COMPARE
ANSI 3-blade 932 10
₹21,100.00INR / Each
COMPARE
ANSI 3-blade 932 18
₹21,600.00INR / Each
COMPARE
ANSI 3-blade 932 10
₹23,600.00INR / Each
COMPARE
ANSI 3-blade 700 2
₹23,800.00INR / Each
COMPARE
ANSI 3-blade 900 4
₹23,600.00INR / Each
COMPARE
ANSI 3-blade 400 10
₹26,200.00INR / Each
Showing 1- 15 of 15