Cole-Parmer® Sanitary Clamps, GL Nylon
High-quality nonmetallic sanitary clamps for critical fluid applications
  • Meet US and EU standards for use in pharmaceutical process applications
  • ASME-BPE, FDA, and USP Class VI compliant
  • Lightweight and reusable
  • Rust and corrosion resistant
  • Compatible with Tri-Clamp® fittings
  • Sterilize by autoclave or gamma irradiation
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
Use the specially designed glass-reinforced nylon sanitary hinged clamps in applications where metal is incompatible. These sanitary clamps provide a leakproof, sterile connection, ensuring the integrity of the fluid being transferred—ideal for pharmaceutical and bioprocessing applications. The universal clamp can also be used with ASME-BPE and BPA standard nonmetallic and stainless flange ferrules.
2,200.00 - ₹5,000.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (in) Description
Availability Pricing
COMPARE
Glass-filled nylon 3/4 Sanitary Clamp, GL Nylon, 3/4"; 1/Ea
₹2,200.00INR / Each
COMPARE
Glass-filled nylon 1-1/2 Sanitary Clamp, GL Nylon, 1-1/2"; 1/Ea
₹2,800.00INR / Each
COMPARE
Glass-filled nylon 2 Sanitary Clamp, GL Nylon, 2"; 1/Ea
₹3,300.00INR / Each
COMPARE
Glass-filled nylon 3 Sanitary Clamp, GL Nylon, 3"; 1/Ea
₹3,900.00INR / Each
COMPARE
Glass-filled nylon 4 Sanitary Clamp, GL Nylon, 4"; 1/Ea
₹5,000.00INR / Each