Cole-Parmer® Spatulas/Macro Spoons, PP
Easily removes residues on flat surfaces
  • Easy to clean
  • Long life cycle
Constructed of flexible polypropylene, these spatula/macro spoon combinations offer excellent collection, mixing, and stirring of solutions, all in one convenient tool. One end features a macro spoon for small, medium, and large size particles and a straight/flat spatula at the other end. The flat end has 35-degree sharp edges for easy removal of the residues on flat surfaces.
Robust and seamless construction guarantees long life cycle. Smooth polished surfaces provide fast and easy cleaning.
3,000.00 - ₹3,800.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Overall Length (in) Width (in)
Availability Pricing
COMPARE
Polypropylene 5 29/32 25/32
₹3,000.00INR / Pkg of 10
COMPARE
Polypropylene 7 1/10 15/16
₹3,600.00INR / Pkg of 10
COMPARE
Polypropylene 8 19/64 1 9/64
₹3,800.00INR / Pkg of 10