Corning CoolRack® Flat-Bottom Tube Cooling Blocks
Cool, freeze, heat, or thaw samples uniformly
  • Place blocks directly into a temperature source from -196°C to over +100°C!
  • Obtain temperature uniformity of ±0.1°C across tubes and plates
  • Quickly reach bath temps—thermoconductive metal construction
These aluminum blocks provide better temperature uniformity than placing samples directly into a bath. Aluminum blocks may be autoclaved, high-heat sterilized, or chemically treated to decrease cross-contamination concerns. Safe for use in ice, dry ice, alcohol, liquid nitrogen, water, oil, and other temperature regulation baths. Use in traditional ice buckets and pans, or use with a Corning CoolBox™ for an ice-free option.
56,400.00 - ₹146,000.00INR / Each
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity Tubes Held Tube Size
Availability Pricing
Corning CoolRack® CF15 Cooling Block, 15 Cryogenic Vials or FACS Tubes; Gray 0439249 Mfr # 432049
COMPARE
15 x 5 mL cryogenic vials or FACS tubes 15
₹56,400.00INR / Each
Corning CoolRack® XT CFT24 Cooling Block, 24 Cryogenic Vials or FACS Tubes; Gray 0439250 Mfr # 432050
COMPARE
24 x 5 mL cryogenic vials or FACS tubes 24
₹107,000.00INR / Each
Corning CoolRack® CF45 Cooling Block, 45 Cryogenic Vials or FACS Tubes; Gray 0439251 Mfr # 432051
COMPARE
45 x 5 mL cryogenic vials or FACS tubes 45
₹106,000.00INR / Each
Corning CoolRack® CFT30 Cooling Block, 30 Cryogenic Vials or FACS Tubes; Gray 0439252 Mfr # 432052
COMPARE
30 x 5 mL cryogenic vials or FACS tubes 30
₹117,000.00INR / Each
Corning CoolRack® 96x0.5mL Cooling Block, 96 x 0.5 mL 2D Tubes; Gray 0439256 Mfr # 432056
COMPARE
96 x 0.5 mL 2D barcoded tubes 96
₹146,000.00INR / Each
Corning CoolRack® 96x1mL Cooling Block, 96 x 1.4 mL 2-D Tubes; Gray 0439257 Mfr # 432057
COMPARE
96 x 1.4 mL 2D barcoded tubes 96
₹133,000.00INR / Each
Corning CoolRack® SV2 Cooling Block, 12 x 5 mL Sample Vials; Gray 0439258 Mfr # 432058
COMPARE
12 x 5 mL sample vials 12
₹122,000.00INR / Each
Corning CoolRack® SV10 Cooling Block, 12 x 10 mL Sample Vials; Gray 0439259 Mfr # 432059
COMPARE
12 x 10 mL sample vials 12
₹122,000.00INR / Each
Corning CoolRack® XT 5mL Cooling Block, 12 x 5 mL Centrifuge Tubes; Gray 0439260 Mfr # 432060
COMPARE
12 x 5 mL centrifuge tubes 12
₹99,500.00INR / Each
Corning CoolRack® 15mL Cooling Block, 9 x 15 mL Centrifuge Tubes; Gray 0439261 Mfr # 432061
COMPARE
9 x 15 mL centrifuge tubes 9
₹104,000.00INR / Each
Corning CoolRack® 50mL Cooling Block, 4 x 50 mL Centrifuge Tubes; Gray 0439262 Mfr # 432062
COMPARE
4 x 50 mL centrifuge tubes 4
₹104,000.00INR / Each
Corning CoolRack® 250mL Cooling Block, 1 x 250 mL Easy-Grip Storage Bottle; Gray 0439264 Mfr # 432064
COMPARE
1 x 250 mL easy-grip storage bottle 1
₹118,000.00INR / Each
Corning CoolRack® V16 Cooling Block, Nine 16x100 mm Tubes; Gray 0439266 Mfr # 432066
COMPARE
9 x 16x100 mm tubes 9
₹108,000.00INR / Each
Corning CoolRack® V13 Cooling Block, Nine 13x100 mm Tubes or 5 mL Cryovials; Gray 0439267 Mfr # 432067
COMPARE
9 x 13x100 mm tubes or 5 mL cryovials 9
₹108,000.00INR / Each
Showing 1- 14 of 14