CPC® (Colder) HFC39 Quick-Disconnect Fittings, Hose Barb Body, Polysulfone with Stainless Steel Valve
Make sterile connections quick and easy with these push-to-connect fittings
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a male coupling insert for easy fluid transfer
  • Audible “click” provides added assurance of a secure connection
  • Quick one-step disconnection process minimize user error
  • 316L stainless steel valve spring stops the flow and offers excellent chemical compatibility
The CPC (Colder) HFC39 series connectors feature an automatic shutoff valve enabling quick disconnection while maintaining its sterility. Connect a male insert and a female body for a secure fluid flow. Simply push the thumb latch to disconnect. A stainless steel valve spring in each connector closes and seals the fluid pathway upon decoupling, protecting the sterility on each side of the system.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
4,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Barb Size / Tubing Id (Port 1) (in) Fitting Material Description
Availability Pricing
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 1/4" ID; 1/Ea 5011222 Mfr # HFCD17439M
COMPARE
1/4 Polysulfone Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 1/4" ID; 1/Ea
₹4,900.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 3/8" ID; 1/Ea 5011224 Mfr # HFCD17639M
COMPARE
3/8 Polysulfone Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 3/8" ID; 1/Ea
₹4,900.00INR / Each
CPC (Colder) HFC39 Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 1/2" ID; 1/Ea 5011226 Mfr # HFCD17839M
COMPARE
1/2 Polysulfone Quick-Disconnect Fitting, Hose Barb Body, Polysulfone with Stainless Steel Valve, 1/2" ID; 1/Ea
₹4,900.00INR / Each