CPC® (Colder) MPC/MPX Quick-Disconnect Fittings, Back-to-Back Insert Adapter
Connect single-use systems that feature identical coupling connections at the end of their tubing
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • Connect to a female coupling body for easy media transfer
  • Combine MPC and MPX couplings for reducing options
The CPC® (Colder) MPC™/MPX® series back-to-back adapters provide an easy way to convert to industry standard connections or single-use systems. Connect a male insert and a female body to create a quick and secure fluid flow. These adapters provide an easy transition between multiple tubing sizes ranging from 1/8" to 1/2" ID. Simply use a cap to seal off the fluid pathway. Coupling halves can rotate when connected reducing tube kinks. Select from various termination size options to suit your application.
All connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, and other critical-use applications.
2,200.00 - ₹4,200.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Connection Type Color
Availability Pricing
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPC Back-to-Back Insert Adapter, Polysulfone; 1/Ea 5011271 Mfr # MPC22C2239M
COMPARE
Polysulfone MPC Insert x MPC Insert Amber tint
₹2,200.00INR / Each
CPC (Colder) MPC™ Quick-Disconnect Fitting, MPC™ to MPX® Back-to-Back Insert Adapter, Polysulfone; 1/Ea 5011272 Mfr # MPC22X2239M
COMPARE
Polysulfone MPC Insert x MPX Insert Amber tint
₹3,100.00INR / Each
CPC (Colder) MPX® Quick-Disconnect Fitting, MPX® to MPX Back-to-Back Insert Adapter, Polysulfone; 1/Ea 5011321 Mfr # MPX22X2239M
COMPARE
Polysulfone MPX Insert x MPX Insert Amber tint
₹4,200.00INR / Each