Digi-Sense 400 Series Thermistor Probes
Thermistor Probes Offer Excellent Accuracy Over the Biological Temperature Range
  • Probes are electronically isolated and come with a 10ft lead with a ¼” phono plug (unless otherwise specified).
  • Probes are designed for multiple applications.
  • Epoxy coated probes can be used in liquid immersions.
  • Time Constant: 1-4 seconds or greater
  • Custom designs or changes to this product are available upon request
Note: Time constant is the time required to reach 63.2% of the new reading. To determine how long it will take to reach 99% of the new reading multiply the time constant by five.
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
2,300.00 - ₹35,300.00INR / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connector Type Max Temperature (° F) Description
Availability Pricing
COMPARE
Phono plug 212 Gen-Purp Flex Thermistor, Vinyl Sheath and Tip, PHono Plug, 10ft Lead
₹13,400.00INR / Each
COMPARE
Phono plug 212 Gen-Purp Flex Thermistor, Vinyl Sheath and Tip, PHono Plug, 100ft Lead
₹25,600.00INR / Each
COMPARE
Phono plug 212 Gen-Purp Flex Thermistor, Vinyl Sheath and Tip, PHono Plug, 50ft Lead
₹27,200.00INR / Each
COMPARE
Phono plug 212 Small Flex Thermistor, PVC Ins Sheath and Tip, PHono Plug, 13ft Lead
₹16,100.00INR / Each
COMPARE
Phono plug 212 Thermistor Probe, 304 Ss, 5 / 32" Dia, 4.5" L, PHono Plug, 10ft Lead
₹22,700.00INR / Each
COMPARE
Phono plug 212 Thermistor Probe, 304 Ss, 1 / 8" Dia, 4.5" L, PHono Plug, 10ft Lead
₹26,900.00INR / Each
COMPARE
Phono plug 212 Air Temperature Probe Epoxy-Encablep 316 SS Cablege 1-7/8 L PHono Plug 10ft Lead
₹25,800.00INR / Each
COMPARE
Phono plug 212 Surface Thermistor, 316 Ss, 3-5 / 8" L, PHono Plug, 10ft Flat Ld
₹26,900.00INR / Each
COMPARE
Phono plug 212 Attach Surface Thermistor, 316 Ss, 1 / 8x3 / 8" , PHono Plug, 10ft Flat Ld
₹19,400.00INR / Each
COMPARE
Phono plug 212 Sm Surf Thermistor, Epoxy Backed, 316 SS, PHono Plug, 24" FEP Leads
₹29,600.00INR / Each
COMPARE
Phono plug 212 Thermistor Probe, Glass, 3 / 16" Dia, 5" L, PHono Plug, 10ft Lead
₹27,500.00INR / Each
COMPARE
Phono plug 212 Pipe-ftg Probe, Ss, 5 / 32" Dia, 1" L, Detachable BNC Conn, PHono Plug
₹35,300.00INR / Each
COMPARE
Stripped Leads 212 Epoxy-Encablepsulated Thermistor Element, 32ga Copper, 3" Bare
₹2,300.00INR / Each
COMPARE
Phono plug 212 Penetration Thermistor 316 SS 5/32" Dia 4-11/16" L PHono Plug 10ft Lead
₹25,300.00INR / Each
COMPARE
Phono plug 212 Sm Flex Thermistor, Nylon and Epoxy Tip, 2.5" L, PHono Plug, 10ft Flat
₹27,800.00INR / Each
Showing 1- 15 of 15