Digi-Sense Thermocouple High-Temperature Wire Probes - Fiberglass Insulated
High-Temperature Fiberglass Insulated Wire Used for High-Temperature Measurements
  • Response Time: 0.75 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type K - yellow, and type J – black..
  • Custom designs or changes to this product are available upon request
Fiberglass wire is not recommended for use in moist or wet environments.
Exposed-junction probes are not recommended for use with aqueous solutions.
Do not subject type J exposed-junction probes to moisture as they will rust.
4,800.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Probe Type Description
Availability Pricing
COMPARE
1300 J Type-J High-Temperature Fiberglass-Insulated Probe Mini Conn Exp 10ft 20-Gauge
₹4,800.00INR / Each
COMPARE
1300 K Type-K High-Temperature Fiberglass-Insulated Probe Mini Conn Exp 10ft 20-Gauge
₹4,800.00INR / Each