DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap
Screw-on cap prevents leakage
  • Include unattached phenolic screw-thread cap
  • Manufactured from borosilicate glass 3.3
  • White marking spot
  • Round-bottom design
  • Autoclavable glass tubes
High-quality reusable culture tubes have uniform wall thickness for maximum heat transfer. Borosilicate glass construction offers excellent chemical resistance. Use the white marking spot to label your samples for easy identification.
117,000.00 - ₹277,000.00INR / Case of 288
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Capacity (mL) Tube Outside Diameter (OD) (mm)
Availability Pricing
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 13 x 100 mm, 8 mL; 288/CS 3459529 Mfr # 45066-13100
COMPARE
Rubber liner 8 13
₹162,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 16 x 100 mm, 12 mL; 288/CS 3459530 Mfr # 45066-16100
COMPARE
Rubber liner 12 16
₹227,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 16 x 125 mm, 16 mL; 288/CS 3459531 Mfr # 45066-16125
COMPARE
Rubber liner 16 16
₹227,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 20 x 125 mm, 25 mL; 192/CS 3459532 Mfr # 45066-20125
COMPARE
Rubber liner 25 20
₹184,000.00INR / Case of 192
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 20 x 150 m, 30 mL; 192/CS 3459533 Mfr # 45066-20150
COMPARE
Rubber liner 30 20
₹210,000.00INR / Case of 192
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 25 x 150 mm, 50 mL; 144/CS 3459534 Mfr # 45066-25150
COMPARE
Rubber liner 50 25
₹173,000.00INR / Case of 144
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, Rubber Liner, 38 x 200 mm, 160 mL; 24/CS 3459535 Mfr # 45066-38200
COMPARE
Rubber liner 160 38
₹117,000.00INR / Case of 24
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 13 x 100 mm, 8 mL; 288/CS 3459536 Mfr # 45066A-13100
COMPARE
PTFE-faced rubber liner 8 13
₹217,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE, 16 x 100 mm, 12 mL; 288/CS 3459537 Mfr # 45066A-16100
COMPARE
PTFE 12 16
₹237,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 16 x 125 mm, 16 mL; 288/CS 3459538 Mfr # 45066A-16125
COMPARE
PTFE-faced rubber liner 16 16
₹261,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE, 16 x 150 mm, 20 mL; 288/CS 3459539 Mfr # 45066A-16150
COMPARE
PTFE 20 16
₹277,000.00INR / Case of 288
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 20 x 125 mm, 25 mL; 192/CS 3459540 Mfr # 45066A-20125
COMPARE
PTFE-faced rubber liner 25 20
₹232,000.00INR / Case of 192
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 20 x 150 mm, 30 mL; 192/CS 3459541 Mfr # 45066A-20150
COMPARE
PTFE-faced rubber liner 30 20
₹244,000.00INR / Case of 192
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 25 x 150 mm, 50 mL; 144/CS 3459542 Mfr # 45066A-25150
COMPARE
PTFE-faced rubber liner 50 25
₹219,000.00INR / Case of 144
DWK Life Sciences (Kimble) KIMAX® Reusable Culture Tubes with Unattached Cap, PTFE-Faced Rubber Liner, 25 x 200 mm, 70 mL; 144/CS 3459543 Mfr # 45066A-25200
COMPARE
PTFE-faced rubber liner 70 25
₹239,000.00INR / Case of 144
Showing 1- 15 of 15