Grieve NTR Series High-Temperature Mechanical Convection Bench Ovens
Constructed for high-temperature use
  • Double doors allow easy loading of baskets and large parts
  • Programmable temperature controller
  • Holds a maximum of 6 shelves
  • 4.3 cu ft capacity
These mechanical convection bench ovens are constructed for high-temperature use. Incoloy® sheathed heating elements and the stainless steel interior are highly resistant to corrosion and oxidation. Industrial rock wool insulation effectively retains heat for improved temperature uniformity. Resilient door gasket contains stainless steel spring inset. Double doors offer greater access to chamber—ideal when loading larger items.
Thermocouple-actuated analog temperature controller provides ±0.5% accuracy. Overtemperature protection turns oven off if unit exceeds maximum temperature; simply press button to reset oven. Red pilot light in front of unit indicates on/off status of heating element.
Models 52300-12 and -14 include two adjustable nickel-plated wire shelves, four inches of insulation. Models 52300-18 and -22 include two adjustable nickel-plated wire shelves and five inches of insulation. The shelves are easily adjustable on 2" centers. The 115 VAC models include a 6-ft power cord with three-prong plug (no plug on 230 VAC models). Order the optional stand separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Max Temperature (° F) Power (VAC)
Availability Pricing
Grieve NTR8002 High-Temperature Mechanical Convection Bench Oven, Max Temp 427 °C (800 °F); 230 V 5230014 Mfr # NTR8002-230V/50HZ
COMPARE
4.3 800 230
Call For Price
Grieve NTR10002 High-Temperature Mechanical Convection Bench Oven, Max Temp 538 °C (1000 °F); 230V 5230022 Mfr # NTR10002/60HZ
COMPARE
4.3 1000 230