IKA® ROTAVISC Complete Viscometer Packages
Stepless measurement of viscosities
  • Four devices for four viscosity ranges
  • Three measuring modes are available for different applications: accurate, balance and fast
  • Ten different programs and ramps can be stored
The ROTAVISC series determines the liquid viscosity in all areas of application ranging from the laboratory to quality control. With the four ROTAVISC models, measure steplessly from 0.01 to 200 rotations per minute. The rotation rate can be set steplessly over the entire measurement range. This allows stepless programming of a flow curve. Two stand options are available. The basic ROTAVISC package comes with the manual ROTASTAND to ensure perpendicular stability. The height of the viscometer can be adjusted steplessly. The step up from this is the HELI package, which comes with the motorized HELISTAND. The HELISTAND measures the viscosity of highly viscous samples with a high level of reliability. It moves the mounted viscometer and its rotating body slowly and uniformly.
1,137,000.00 - ₹1,416,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description Viscosity
Availability Pricing
IKA ROTAVISC lo-vi HELI Complete Viscometer Package 8305537 Mfr # 25008214
COMPARE
ROTAVISC lo-vi HELI Complete Viscometer Package 6,000,000 mPa-s
₹1,416,000.00INR / Each
IKA ROTAVISC me-vi HELI Complete Viscometer Package 8305538 Mfr # 25008216
COMPARE
ROTAVISC me-vi HELI Complete Viscometer Package 40,000,000 mPa-s
₹1,416,000.00INR / Each
IKA ROTAVISC hi-vi I HELI Complete Viscometer Package 8305539 Mfr # 25008217
COMPARE
ROTAVISC hi-vi I HELI Complete Viscometer Package 80,000,000 mPa-s
₹1,416,000.00INR / Each
IKA ROTAVISC hi-vi II HELI Complete Viscometer Package 8305545 Mfr # 0010013765
COMPARE
ROTAVISC hi-vi II HELI Complete Viscometer Package 320,000,000 mPa-s
₹1,416,000.00INR / Each
IKA ROTAVISC me-vi Complete Viscometer Package 8305547 Mfr # 25000311
COMPARE
ROTAVISC me-vi Complete Viscometer Package 40,000,000 mPa-s
₹1,137,000.00INR / Each
IKA ROTAVISC hi-vi I Complete Viscometer Package 8305548 Mfr # 25000312
COMPARE
ROTAVISC hi-vi I Complete Viscometer Package 80,000,000 mPa-s
₹1,137,000.00INR / Each
IKA ROTAVISC hi-vi II Complete Viscometer Package 8305549 Mfr # 25000313
COMPARE
ROTAVISC hi-vi II Complete Viscometer Package 320,000,000 mPa-s
₹1,137,000.00INR / Each
Showing 1- 7 of 7