IKA T-25 Homogenizer Rotor-Stator Generators
Select a generator according to your sample volume, working solution, and needed dispersion type. The standard-capacity generator is available in three volume ranges. The large-capacity generator is available in both coarse (20 ìm) and fine (5 ìm) dispersion.
35,300.00 - ₹178,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Dispersion Type Max Sample Size (mL) Min Sample Size (mL)
Availability Pricing
IKA 1713300 Homogenizer Rotor-Stator Generator, Coarse, 200mm x 25mm, 50-2500 mL 0471952 Mfr # 0001713300
₹149,000.00INR / Each
COMPARE
Coarse 2000 50
Loading
₹149,000.00INR / Each
IKA 1713800 Homogenizer Rotor-Stator Generator, Fine, 195mm x 25mm, 100-2000 mL 0471953 Mfr # 0001713800
₹166,000.00INR / Each
COMPARE
Fine 2000 100
Loading
₹166,000.00INR / Each
IKA 0594000 Homogenizer Rotor-Stator Generator, Coarse, 105mm x 10mm, 1-100 mL 0472010 Mfr # 0000594000
₹121,000.00INR / Each
COMPARE
Coarse 100 1
Loading
₹121,000.00INR / Each
IKA 1024200 Homogenizer Rotor-Stator Generator, Coarse, 108mm x 8mm, 1-50 mL 0472011 Mfr # 0001024200
₹178,000.00INR / Each
COMPARE
Coarse 50 1
Loading
₹178,000.00INR / Each
IKA 0593400 Homogenizer Rotor-Stator Generator, Coarse, 200mm x 18mm, 10-1500 mL 0472012 Mfr # 0000593400
₹114,000.00INR / Each
COMPARE
Coarse 1500 10
Loading
₹114,000.00INR / Each
IKA 3452200 Plastic Dispersing Element 10mm X 150mm (10/pk) For T-25 Digital Homogenizer 0472091 Mfr # 0003452200
₹35,300.00INR / Pkg of 10
COMPARE
Loading
₹35,300.00INR / Pkg of 10