Kimax, Serialized and Certified, Class A Burette (Buret)
Get more accurate delivery
  • Fill burette (buret) easily through PTFE stopcock with 1⁄4" ID flexible tubing
Designed from ASTM E287-Class A requirements, this self-zeroing burette (buret) is permanantly marked with an individual serial number. Filling is completed thru the (insert image P/S) stopcock and accepts 1/4" ID flexible tubing for filling tube and overflow tube. Graduations are in black enamel.
89,100.00 - ₹107,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Graduation Interval (mL) Tolerance
Availability Pricing
DWK Life Sciences (Kimble) Kimax Serialized and Certified Class A Glass Burette, 10 mL; 1/Pk 3454100 Mfr # 17054F-10
COMPARE
10 0.05 0.02
₹89,100.00INR / Each
DWK Life Sciences (Kimble) Kimax Serialized and Certified Class A Glass Burette, 25 mL; 1/Pk 3454102 Mfr # 17054F-25
COMPARE
25 0.1 0.03
₹89,200.00INR / Each
DWK Life Sciences (Kimble) Kimax Serialized and Certified Class A Glass Burette, 50 mL; 1/Pk 3454104 Mfr # 17054F-50
COMPARE
50 0.1 0.02
₹92,100.00INR / Each
DWK Life Sciences (Kimble) Kimax Serialized and Certified Class A Glass Burette, 100 mL; 1/Pk 3454106 Mfr # 17054F-100
COMPARE
100 0.2 0.1
₹107,000.00INR / Each